Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021492.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021492.hg.1 |
ENST00000543122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021492.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021492.hg.1 |
TC17001597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010878.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
ENST00000331780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
ENST00000586359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
ENST00000588866.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
NM_152343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187247.hg.1 |
TC1700010878.hg.1 |
Clariom D TC |