Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014246.1 | NP_055061.1 | 11 - 12 | 5526 | 70 | |
XM_006724383.3 | XP_006724446.1 | 11 - 12 | 5853 | 70 | ||
XM_011530553.1 | XP_011528855.1 | 11 - 12 | 5858 | 70 | ||
XM_011530554.2 | XP_011528856.1 | 11 - 12 | 2069 | 70 | ||
XM_011530555.2 | XP_011528857.1 | 11 - 12 | 2150 | 70 | ||
GenBank mRNA | AF231024.1 | - | 11 - 12 | 5526 | 70 | |
AK302517.1 | - | 3 - 4 | 604 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |