Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKIPSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016453.3 | NP_057537.1 | 3 - 4 | 612 | 78 | |
XM_017006594.1 | XP_016862083.1 | 3 - 4 | 606 | 78 | ||
XM_017006596.1 | XP_016862085.1 | 3 - 4 | 626 | 78 | ||
XM_017006597.1 | XP_016862086.1 | 3 - 4 | 370 | 78 | ||
XM_017006599.1 | XP_016862088.1 | 3 - 4 | 632 | 78 | ||
GenBank mRNA | AF178432.1 | - | 3 - 4 | 580 | 78 | |
AF303581.1 | - | 3 - 4 | 551 | 78 | ||
AJ242655.2 | - | 3 - 4 | 580 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |