Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142449.1 | NP_001135921.1 | 5 - 6 | 1119 | 60 | |
NM_001142451.1 | NP_001135923.1 | 6 - 7 | 1185 | 60 | ||
XM_006721096.3 | XP_006721159.1 | 7 - 8 | 955 | 60 | ||
GenBank mRNA | AL390215.1 | - | 5 - 6 | 1081 | 60 | |
BC047741.1 | - | 6 - 7 | 1159 | 5757339 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons |