Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000262953.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000426948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000587217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000587672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000589205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000589291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000590101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000590183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000590536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000591529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000592584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
NM_001144761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
NM_001300846.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
NM_003260.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
TC1900009269.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000262953.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000426948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000587217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000587672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000589205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000589291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000590101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000590183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000590536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000591529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
ENST00000592584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
NM_001144761.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
NM_001300846.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900173855.hg.1 |
NM_003260.4 |
Refseq |