Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00948386_m1
See other FHAD1 GE Assays ›
Gene Symbol:
FHAD1
Gene Name:
forkhead associated phosphopeptide binding domain 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 15236559 - 15400283 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00948386_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: FHAD1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_011540575.2
XP_011538877.1
14 - 15
2120
98
XM_011540576.2
XP_011538878.1
14 - 15
2121
98
XM_011540577.2
XP_011538879.1
14 - 15
2082
98
XM_011540580.2
XP_011538882.1
14 - 15
2119
98
XM_011540581.2
XP_011538883.1
14 - 15
2119
98
XM_011540582.2
XP_011538884.1
14 - 15
2119
98
XM_011540584.2
XP_011538886.1
14 - 15
2120
98
XM_011540588.2
XP_011538890.1
14 - 15
2111
98
XM_011540589.2
XP_011538891.1
14 - 15
2120
98
XM_011540590.2
XP_011538892.1
14 - 15
2112
98
XM_011540592.2
XP_011538894.1
13 - 14
1821
98
XM_011540593.2
XP_011538895.1
14 - 15
2116
98
XM_011540595.2
XP_011538897.1
11 - 12
1464
98
XM_011540596.2
XP_011538898.1
8 - 9
1024
98
XM_011540597.2
XP_011538899.1
14 - 15
2121
98
XM_017000198.1
XP_016855687.1
14 - 15
2121
98
XM_017000199.1
XP_016855688.1
14 - 15
2121
98
XM_017000200.1
XP_016855689.1
14 - 15
2120
98
XM_017000201.1
XP_016855690.1
14 - 15
2120
98
XM_017000202.1
XP_016855691.1
13 - 14
2013
98
XM_017000203.1
XP_016855692.1
14 - 15
2117
98
XM_017000204.1
XP_016855693.1
14 - 15
2116
98
XM_017000205.1
XP_016855694.1
12 - 13
1926
98
XM_017000206.1
XP_016855695.1
14 - 15
2116
98
XM_017000207.1
XP_016855696.1
13 - 14
1885
98
XM_017000208.1
XP_016855697.1
14 - 15
2111
98
XM_017000209.1
XP_016855698.1
14 - 15
2115
98
XM_017000210.1
XP_016855699.1
14 - 15
2111
98
XM_017000211.1
XP_016855700.1
14 - 15
2119
98
XM_017000212.1
XP_016855701.1
14 - 15
2110
98
XM_017000213.1
XP_016855702.1
14 - 15
2120
98
XM_017000214.1
XP_016855703.1
14 - 15
2121
98
GenBank mRNA
AK057963.1
-
12 - 13
1754
98
AL833342.1
-
3 - 4
372
98
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Homo sapiens
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0