Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027539.hg.1 |
TC07001761.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027539.hg.1 |
ENST00000342288.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027539.hg.1 |
ENST00000352877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027539.hg.1 |
ENST00000428084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027539.hg.1 |
ENST00000437633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027539.hg.1 |
ENST00000494651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07027539.hg.1 |
NM_014705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000417165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000423057.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000428053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000428084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000437129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000437633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000445943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000450156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000464338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000492436.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
NM_014705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000417165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000423057.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000428053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000428084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000437129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000437633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000445943.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000450156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000464338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
ENST00000492436.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
NM_014705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700188533.hg.1 |
TC0700012276.hg.1 |
Clariom D TC |