Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014034.hg.1 |
TC01000891.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014034.hg.1 |
ENST00000263178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014034.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014034.hg.1 |
ENST00000370185.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014034.hg.1 |
ENST00000457765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014034.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014034.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165397.hg.1 |
TC0100009181.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000370185.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
ENST00000457765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_001166252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009181.hg.1 |
NM_014839.4 |
Refseq |