Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003299.hg.1 |
ENST00000081029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003299.hg.1 |
ENST00000321446.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003299.hg.1 |
ENST00000538315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003299.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003299.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003299.hg.1 |
TC12000270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003300.hg.1 |
ENST00000081029.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003300.hg.1 |
ENST00000321446.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003300.hg.1 |
ENST00000538315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003300.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003300.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12003300.hg.1 |
TC12000270.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150302.hg.1 |
TC1200007161.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200150300.hg.1 |
TC1200007161.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000081029.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000536569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000538315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
ENST00000542791.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_001190864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007161.hg.1 |
NM_021821.3 |
Refseq |