Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013663.hg.1 |
TC07000876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013663.hg.1 |
ENST00000422142.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013663.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013664.hg.1 |
TC07000876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013664.hg.1 |
ENST00000422142.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013664.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013668.hg.1 |
TC07000876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013668.hg.1 |
ENST00000422142.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013668.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013665.hg.1 |
TC07000876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013665.hg.1 |
ENST00000422142.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013665.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013667.hg.1 |
TC07000876.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013667.hg.1 |
ENST00000422142.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013667.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166371.hg.1 |
TC0700009346.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009346.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166368.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166368.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166368.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166368.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166368.hg.1 |
TC0700009346.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
ENST00000520413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166370.hg.1 |
TC0700009346.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700166366.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166366.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166366.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166366.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166366.hg.1 |
TC0700009346.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700166367.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166367.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166367.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166367.hg.1 |
ENST00000422142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166367.hg.1 |
ENST00000521281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700166367.hg.1 |
NM_001080511.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700166367.hg.1 |
TC0700009346.hg.1 |
Clariom D TC |