Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017008866.1 | XP_016864355.1 | 1 - 2 | 214 | 59 | |
XM_017008868.1 | XP_016864357.1 | 1 - 2 | 214 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |