Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016642.hg.1 |
ENST00000418657.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
ENST00000291890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
ENST00000338835.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
ENST00000350790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
ENST00000357397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
ENST00000447255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
NM_001145457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
NM_001145458.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
NM_001242356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
NM_001242357.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
NM_004829.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012273.hg.1 |
TC19000886.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
ENST00000291890.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
ENST00000338835.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
ENST00000350790.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
ENST00000447255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
NM_001145457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
NM_001145458.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
NM_001242356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
NM_001242357.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
NM_004829.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012274.hg.1 |
TC19000886.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008882.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
TC1900008882.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000357397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000598576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169505.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
TC1900008882.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000291890.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000338835.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000350790.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000594765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000601137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169507.hg.1 |
ENST00000601903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016642.hg.1 |
ENST00000418657.2 |
Ensembl |