Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020728.2 | NP_065779.1 | 11 - 12 | 1449 | 142 | |
GenBank mRNA | AB033054.2 | - | 12 - 13 | 1235 | 142 | |
AK126214.1 | - | 11 - 12 | 1094 | 142 | ||
AK308824.1 | - | 11 - 12 | 1057 | 142 | ||
AL833233.1 | - | 11 - 12 | 1058 | 142 | ||
AY368150.1 | - | 11 - 12 | 1449 | 142 | ||
BC013957.2 | - | 2 - 3 | 108 | 142 | ||
DQ993201.1 | - | 13 - 14 | 1467 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |