Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008996.hg.1 |
ENST00000256010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008996.hg.1 |
ENST00000551529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008996.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008996.hg.1 |
TC12000687.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008995.hg.1 |
ENST00000256010.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008995.hg.1 |
ENST00000551529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008995.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008995.hg.1 |
TC12000687.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200161421.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161421.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161421.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161421.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161421.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161421.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161421.hg.1 |
TC1200008353.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161420.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161420.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161420.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161420.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161420.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161420.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161420.hg.1 |
TC1200008353.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200161422.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161422.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161422.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161422.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161422.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200161422.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200161422.hg.1 |
TC1200008353.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000256010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000550879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
ENST00000551529.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008353.hg.1 |
NM_006183.4 |
Refseq |