Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005437.hg.1 |
TC02000362.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005437.hg.1 |
ENST00000282549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005437.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005440.hg.1 |
TC02000362.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005440.hg.1 |
ENST00000282549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005440.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000366671.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_001199770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007768.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153609.hg.1 |
TC0200007768.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000282549.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000405984.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000476383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000477348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
ENST00000484066.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
NM_014562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
NR_130153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200153613.hg.1 |
TC0200007768.hg.1 |
Clariom D TC |