Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014602.hg.1 |
TC08001138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014602.hg.1 |
ENST00000287263.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014602.hg.1 |
ENST00000330843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014602.hg.1 |
ENST00000343853.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014602.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014602.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014596.hg.1 |
TC08001138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014596.hg.1 |
ENST00000330843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014596.hg.1 |
ENST00000523182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014596.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014603.hg.1 |
TC08001138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014603.hg.1 |
ENST00000287263.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014603.hg.1 |
ENST00000330843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014603.hg.1 |
ENST00000343853.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014603.hg.1 |
ENST00000522727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014603.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014603.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014598.hg.1 |
TC08001138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014598.hg.1 |
ENST00000287263.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014598.hg.1 |
ENST00000330843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014598.hg.1 |
ENST00000522727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014598.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014598.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014598.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014606.hg.1 |
TC08001138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014606.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014606.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014599.hg.1 |
TC08001138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014599.hg.1 |
ENST00000287263.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014599.hg.1 |
ENST00000330843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014599.hg.1 |
ENST00000522727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014599.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014599.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014599.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168932.hg.1 |
TC0800010138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168924.hg.1 |
TC0800010138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800168936.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168936.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168936.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168936.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168936.hg.1 |
TC0800010138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168927.hg.1 |
TC0800010138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168930.hg.1 |
TC0800010138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000522774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000523182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
ENST00000524118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010138.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
ENST00000287263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
ENST00000330843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
ENST00000522727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
NM_001002814.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
NM_025151.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800168933.hg.1 |
TC0800010138.hg.1 |
Clariom D TC |