Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080114.1 | NP_001073583.1 | 3 - 4 | 343 | 78 | |
NM_001080116.1 | NP_001073585.1 | 3 - 4 | 448 | 78 | ||
XM_011539189.1 | XP_011537491.1 | 4 - 5 | 407 | 78 | ||
XM_011539190.1 | XP_011537492.1 | 3 - 4 | 343 | 78 | ||
XM_017015609.1 | XP_016871098.1 | 4 - 5 | 407 | 78 | ||
GenBank mRNA | AJ133766.1 | - | 3 - 4 | 399 | 78 | |
AJ133767.1 | - | 3 - 4 | 320 | 78 | ||
AK300692.1 | - | 4 - 5 | 356 | 78 | ||
AL601484.1 | - | 3 - 4 | 355 | 78 | ||
BC010929.2 | - | 3 - 4 | 345 | 78 | ||
DA898241.1 | - | 3 - 4 | 448 | 78 | ||
KU178543.1 | - | 3 - 4 | 320 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |