Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017177.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017177.hg.1 |
TC14001528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017178.hg.1 |
ENST00000216756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017178.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017178.hg.1 |
TC14001528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017176.hg.1 |
ENST00000216756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017176.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017176.hg.1 |
TC14001528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017169.hg.1 |
ENST00000216756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017169.hg.1 |
ENST00000536961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017169.hg.1 |
ENST00000541568.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017169.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017169.hg.1 |
TC14001528.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010263.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000216756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000536961.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000541568.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000558523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000558764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000559504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000559514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
ENST00000561468.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
NM_032630.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400175946.hg.1 |
TC1400010263.hg.1 |
Clariom D TC |