Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007222.hg.1 |
ENST00000631190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007222.hg.1 |
ENST00000631190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007222.hg.1 |
NR_015451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007222.hg.1 |
NR_015451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010481.hg.1 |
ENST00000628571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010481.hg.1 |
ENST00000628571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010482.hg.1 |
ENST00000527583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000554634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009601.hg.1 |
ENST00000555771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007222.hg.1 |
ENST00000631190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007222.hg.1 |
ENST00000631190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007222.hg.1 |
NR_015451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007222.hg.1 |
NR_015451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010482.hg.1 |
ENST00000527583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010481.hg.1 |
ENST00000628571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010481.hg.1 |
ENST00000628571.1 |
Ensembl |