Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001426.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001426.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001426.hg.1 |
TC18000127.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007013.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800148023.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148023.hg.1 |
NM_001944.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800148023.hg.1 |
TC1800007013.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800148023.hg.1 |
ENST00000257189.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148023.hg.1 |
NM_001944.2 |
Refseq |