Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012706.hg.1 |
TC08001012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012706.hg.1 |
ENST00000180173.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012706.hg.1 |
ENST00000521857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012706.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012706.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012704.hg.1 |
TC08001012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012704.hg.1 |
ENST00000180173.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012704.hg.1 |
ENST00000521857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012704.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08012704.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166097.hg.1 |
TC0800009726.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166099.hg.1 |
TC0800009726.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000180173.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000517317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000518272.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519122.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000519763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000521857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
ENST00000523571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009726.hg.1 |
NM_004686.4 |
Refseq |