Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282569.1 | NP_001269498.1 | 3 - 4 | 286 | 109 | |
NM_001282573.1 | NP_001269502.1 | 3 - 4 | 286 | 109 | ||
XM_006710681.3 | XP_006710744.1 | 3 - 4 | 360 | 109 | ||
GenBank mRNA | AK302248.1 | - | 3 - 4 | 236 | 109 | |
DA737244.1 | - | 3 - 4 | 236 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |