Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133269.3 | NP_579803.1 | 4 - 5 | 775 | 66 | |
NM_133278.3 | NP_579812.1 | 3 - 4 | 739 | 66 | ||
GenBank mRNA | D87853.1 | - | 4 - 5 | 578 | 66 | |
D87859.1 | - | 5 - 6 | 656 | 66 | ||
D87862.1 | - | 3 - 4 | 542 | 66 | ||
U43774.1 | - | 4 - 5 | 583 | 66 | ||
X92559.1 | - | 3 - 4 | 542 | 66 | ||
X92560.1 | - | 4 - 5 | 578 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |