Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000551604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000551604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
ENST00000551604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011350.hg.1 |
ENST00000261726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011350.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011350.hg.1 |
TC12000863.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200165761.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200165761.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200165761.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200165761.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200165761.hg.1 |
TC1200008860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000261726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000397643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000551604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000551604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
ENST00000551604.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008860.hg.1 |
NM_015267.3 |
Refseq |