Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000342704.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000344201.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000346541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000375279.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000397800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000471007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000471007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000471007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000471007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000471007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000471007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000491618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000491618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000491618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000491618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000491618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000491618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000492345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000543126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000543126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000543126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000543126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000543126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
ENST00000543126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001032999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_001039709.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007157.hg.1 |
NM_005093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149079.hg.1 |
ENST00000359606.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149079.hg.1 |
TC2000007157.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149079.hg.1 |
ENST00000359606.3 |
Ensembl |