Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007752.hg.1 |
ENST00000360321.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007752.hg.1 |
ENST00000382369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007752.hg.1 |
ENST00000400269.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007752.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007752.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20007752.hg.1 |
TC20000538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
TC2000008121.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000156994.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000360321.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000382369.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
ENST00000400269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_080571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008121.hg.1 |
NM_153269.2 |
Refseq |