Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDS5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100399.1 | NP_001093869.1 | 29 - 30 | 3945 | 129 | |
XM_011513672.2 | XP_011511974.1 | 29 - 30 | 3558 | 129 | ||
XM_017007928.1 | XP_016863417.1 | 29 - 30 | 3520 | 129 | ||
GenBank mRNA | AB014548.1 | - | 17 - 18 | 1949 | 129 | |
AB621803.1 | - | 29 - 30 | 3959 | 129 | ||
AF294791.1 | - | 29 - 30 | 3516 | 129 | ||
AK021757.1 | - | 4 - 5 | 431 | 129 | ||
AK023592.1 | - | 2 - 3 | 198 | 129 | ||
BC009650.1 | - | 5 - 6 | 530 | 129 | ||
BC114218.1 | - | 29 - 30 | 3641 | 5579832 | 129 | |
BC126225.1 | - | 28 - 29 | 3355 | 8991941 | 129 | |
CR749425.1 | - | 28 - 29 | 3314 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |