Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006416.4 | NP_006407.1 | 6 - 7 | 835 | 115 | |
GenBank mRNA | AK293696.1 | - | 5 - 6 | 627 | 115 | |
AL122119.1 | - | 5 - 6 | 763 | 115 | ||
BC017807.1 | - | 6 - 7 | 783 | 4690513 | 115 | |
D87969.1 | - | 6 - 7 | 783 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |