Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014611.2 | NP_055426.1 | 71 - 72 | 12017 | 87 | |
XM_005248700.3 | XP_005248757.1 | 32 - 33 | 5949 | 87 | ||
XM_006715405.3 | XP_006715468.1 | 35 - 36 | 7495 | 87 | ||
XM_011535635.2 | XP_011533937.1 | 71 - 72 | 11822 | 87 | ||
XM_011535636.2 | XP_011533938.1 | 71 - 72 | 11987 | 87 | ||
GenBank mRNA | AB002299.2 | - | 25 - 26 | 4673 | 87 | |
AF503925.1 | - | 71 - 72 | 11946 | 87 | ||
AK125181.1 | - | 9 - 10 | 1817 | 87 | ||
CR627399.1 | - | 17 - 18 | 3472 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |