Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005246.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005246.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005246.hg.1 |
TC20000363.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000152442.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152442.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152442.hg.1 |
TC2000007514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000152442.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152442.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
ENST00000372330.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007514.hg.1 |
NM_004994.2 |
Refseq |