Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021208.hg.1 |
ENST00000221954.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021208.hg.1 |
NM_001817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021208.hg.1 |
TC19001559.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000221954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000472081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010761.hg.1 |
ENST00000600925.1 |
Ensembl |