Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
ENST00000281171.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
ENST00000348962.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
ENST00000442921.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
ENST00000445537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
ENST00000542557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
ENST00000544244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
NM_002848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
NM_030667.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
NM_030668.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
NM_030669.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
NM_030670.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
NM_030671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002519.hg.1 |
TC12000203.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015356.hg.1 |
ENST00000422899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015356.hg.1 |
ENST00000422899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015356.hg.1 |
ENST00000422899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015356.hg.1 |
ENST00000448977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015356.hg.1 |
ENST00000448977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007201.hg.1 |
ENST00000631361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007201.hg.1 |
ENST00000631361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015356.hg.1 |
ENST00000422899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015356.hg.1 |
ENST00000422899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015356.hg.1 |
ENST00000422899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015356.hg.1 |
ENST00000448977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015356.hg.1 |
ENST00000448977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000281171.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000348962.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000442921.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000445537.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000535311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000538907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000542557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000544244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_002848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030667.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030668.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000281171.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000348962.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000442921.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000445537.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000535311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000538907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000542557.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
ENST00000544244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_002848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030667.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030668.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030669.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030670.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
NM_030671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148646.hg.1 |
TC1200006957.hg.1 |
Clariom D TC |