Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000517358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000517358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027552.hg.1 |
TC05003434.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027552.hg.1 |
ENST00000307851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027552.hg.1 |
ENST00000522593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027552.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197274.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500197274.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500197274.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197274.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500197274.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500197274.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197274.hg.1 |
TC0500013388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000307851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000517358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000517358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000521665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
ENST00000522902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013388.hg.1 |
NM_032782.4 |
Refseq |