Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002919.hg.1 |
ENST00000358821.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002919.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002919.hg.1 |
TC18000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000578498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000578498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000578498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
TC1800007682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152584.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000358821.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000578498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000578498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000578498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000582461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584004.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000584316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
ENST00000585136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007682.hg.1 |
NM_032649.5 |
Refseq |