Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000317060.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
ENST00000585548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007113.hg.1 |
NM_152654.2 |
Refseq |