Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014404.hg.1 |
TC09002889.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014404.hg.1 |
ENST00000432825.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014404.hg.1 |
ENST00000541829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014404.hg.1 |
ENST00000543968.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014404.hg.1 |
ENST00000544475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09014404.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185919.hg.1 |
TC0900012227.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000276960.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000432825.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000541607.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000543968.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
ENST00000544208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012227.hg.1 |
NM_012166.2 |
Refseq |