Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006287.hg.1 |
ENST00000347785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006287.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006287.hg.1 |
TC11000526.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007790.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155173.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155173.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155173.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155173.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155173.hg.1 |
ENST00000544014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155173.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155173.hg.1 |
TC1100007790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100155174.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155174.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155174.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155174.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155174.hg.1 |
TC1100007790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100155172.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155172.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155172.hg.1 |
ENST00000347785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155172.hg.1 |
NM_014207.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155172.hg.1 |
TC1100007790.hg.1 |
Clariom D TC |