Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |