Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041438.hg.1 |
TC02002582.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041438.hg.1 |
ENST00000295749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041438.hg.1 |
ENST00000410053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041438.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041440.hg.1 |
TC02002582.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041440.hg.1 |
ENST00000410053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041440.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041437.hg.1 |
TC02002582.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041437.hg.1 |
ENST00000295749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041437.hg.1 |
ENST00000410053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041437.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041439.hg.1 |
TC02002582.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041439.hg.1 |
ENST00000295749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041439.hg.1 |
ENST00000410053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041439.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210699.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210699.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210699.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210699.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210699.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210699.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210699.hg.1 |
TC0200015099.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200210703.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210703.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210703.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210703.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210703.hg.1 |
TC0200015099.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200210700.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210700.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210700.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210700.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210700.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210700.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210700.hg.1 |
TC0200015099.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200210702.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210702.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210702.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210702.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210702.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200210702.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200210702.hg.1 |
TC0200015099.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000404136.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
ENST00000410053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015099.hg.1 |
NM_020943.2 |
Refseq |