Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039975.hg.1 |
TC01002573.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039975.hg.1 |
ENST00000296387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039975.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039975.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039975.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203844.hg.1 |
TC0100013896.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000296387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000372539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
ENST00000539749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001123395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_001185117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013896.hg.1 |
NM_148960.2 |
Refseq |