Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000317.hg.1 |
TC6_qbl_hap6000026.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000317.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000313.hg.1 |
TC6_ssto_hap7000026.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000313.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000231.hg.1 |
TC6_apd_hap1000025.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000231.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002768.hg.1 |
TC06000334.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002768.hg.1 |
ENST00000376694.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002768.hg.1 |
ENST00000376695.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002768.hg.1 |
ENST00000498045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002768.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000321.hg.1 |
TC6_dbb_hap3000026.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000321.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000336.hg.1 |
TC6_cox_hap2000033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000336.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000312.hg.1 |
TC6_mann_hap4000028.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000312.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376688.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376688.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000295.hg.1 |
TC6_mcf_hap5000022.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000295.hg.1 |
NM_033229.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376688.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376688.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007512.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000376694.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000433744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000477944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
ENST00000619857.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600150821.hg.1 |
TC0600007512.hg.1 |
Clariom D TC |