Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009424.hg.1 |
TC08000698.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009424.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009424.hg.1 |
ENST00000523492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009424.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009423.hg.1 |
TC08000698.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009423.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009423.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159431.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159431.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159431.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159431.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159431.hg.1 |
TC0800008674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000518057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008674.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
ENST00000286234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
ENST00000523492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
NM_001283012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
NM_022783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159432.hg.1 |
TC0800008674.hg.1 |
Clariom D TC |