Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000476813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000476813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000476813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003082.hg.1 |
TC01000188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003082.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003082.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375943.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000476813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000476813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000476813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
ENST00000483406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006983.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148650.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148650.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148650.hg.1 |
ENST00000375949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148650.hg.1 |
NM_007272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148650.hg.1 |
TC0100006983.hg.1 |
Clariom D TC |