Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000492154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000492154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
ENST00000492154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009976.hg.1 |
TC02000627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009976.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009976.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009975.hg.1 |
TC02000627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009975.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009975.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160680.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160680.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160680.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160680.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160680.hg.1 |
TC0200008679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200160679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160679.hg.1 |
TC0200008679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000295269.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000492154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000492154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
ENST00000492154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008679.hg.1 |
NM_001011552.3 |
Refseq |