Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000466755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000466755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000466755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000478845.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001706.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001706.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001706.hg.1 |
TC15000270.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500147962.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147962.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147962.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147962.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147962.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147962.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147962.hg.1 |
TC1500006925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000260356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000397591.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000466755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000466755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000466755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000478845.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000484734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000490247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000497720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000559746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
ENST00000560894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006925.hg.1 |
NM_003246.3 |
Refseq |