Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056375.hg.1 |
TC01003680.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056375.hg.1 |
ENST00000331314.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056375.hg.1 |
ENST00000367346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056375.hg.1 |
ENST00000367348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056375.hg.1 |
ENST00000447706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056375.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056373.hg.1 |
TC01003680.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056373.hg.1 |
ENST00000331314.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056373.hg.1 |
ENST00000367346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056373.hg.1 |
ENST00000367348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056373.hg.1 |
ENST00000447706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056373.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000436897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000436897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000452560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000452560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000452560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056374.hg.1 |
TC01003680.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056374.hg.1 |
ENST00000331314.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056374.hg.1 |
ENST00000367346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056374.hg.1 |
ENST00000367348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056374.hg.1 |
ENST00000447706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056374.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227762.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227762.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227762.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227762.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227762.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227762.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227762.hg.1 |
TC0100016893.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000413408.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000433235.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000436897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000436897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000452560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000452560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000452560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
ENST00000453944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016893.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
ENST00000429498.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227761.hg.1 |
TC0100016893.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100227763.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227763.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227763.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227763.hg.1 |
ENST00000331314.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227763.hg.1 |
ENST00000447706.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227763.hg.1 |
NM_001031725.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227763.hg.1 |
TC0100016893.hg.1 |
Clariom D TC |