Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025513.hg.1 |
TC03001422.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025513.hg.1 |
ENST00000296473.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025513.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025513.hg.1 |
ENST00000483022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025513.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025512.hg.1 |
TC03001422.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025512.hg.1 |
ENST00000296473.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025512.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03025512.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182439.hg.1 |
TC0300011089.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000455683.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000473451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000486107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
ENST00000493206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_001142501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011089.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000296473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000417270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000483022.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
ENST00000484985.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
NM_032355.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300182440.hg.1 |
TC0300011089.hg.1 |
Clariom D TC |