Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006178.hg.1 |
TC02004969.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006178.hg.1 |
ENST00000244227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006178.hg.1 |
ENST00000409116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006178.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006178.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006179.hg.1 |
TC02004969.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006179.hg.1 |
ENST00000244227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006179.hg.1 |
ENST00000409116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006179.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006179.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006180.hg.1 |
TC02004969.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006180.hg.1 |
ENST00000244227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006180.hg.1 |
ENST00000409116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006180.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006180.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222218.hg.1 |
TC0200016470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
ENST00000488986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016470.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222219.hg.1 |
TC0200016470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000244227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000409116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000450162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
ENST00000487029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
NM_006857.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
NR_037862.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222217.hg.1 |
TC0200016470.hg.1 |
Clariom D TC |