Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005464.hg.1 |
ENST00000325718.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005464.hg.1 |
ENST00000484263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005464.hg.1 |
NM_005937.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005464.hg.1 |
TC17000462.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000612066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000612066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000612066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000611305.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000612066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000612066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000612066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000614769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000615858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000618652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000619356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620482.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000620609.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007716.hg.1 |
ENST00000621332.4 |
Ensembl |