Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000338641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000361166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000361676.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000397789.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000397822.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000403435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000403999.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
ENST00000413209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
NM_000268.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
NM_016418.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
NM_181825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
NM_181828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
NM_181832.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
NM_181833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003702.hg.1 |
TC22000199.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
TC2200007027.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000338641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000361166.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000361676.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000397789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000403435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000403999.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000413209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_000268.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_016418.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000338641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000361166.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000361676.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000397789.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000403435.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000403999.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
ENST00000413209.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_000268.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_016418.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200149113.hg.1 |
NM_181833.2 |
Refseq |